N(α)-Acetyl-D-tryptophan

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-D-Trp-OH, Ac-D-Trp-OH, N-Acetyl-D-tryptophan

4027317.0005 4027317.0025
BACE4027317.0005EA 0 EUR
BACE4027317.0005 BACE4027317.0025
N(α)-Acetyl-D-tryptophan
N(α)-Acetyl-D-tryptophan
Formule: C₁₃H₁₄N₂O₃
MW: 246.3 g/mol
MDL Nummer: MFCD00065021
CAS nummer: 2280-01-5
EINECS: 218-912-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR