Tetrahydrofurfuryl acetate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Tetrahydrofurfuryl Acetate
T0105-25G
T0105-500G
TCIAT0105-25GEA
55
EUR
TCIAT0105-25G
TCIAT0105-500G
Tetrahydrofurfuryl acetate ≥97.0% (by GC)
Tetrahydrofurfuryl acetate
Formule:
C₇H₁₂O₃ Vlampunt: 84°C |
MDL Nummer:
MFCD00022478 CAS nummer: 637-64-9 EINECS: 211-296-8 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...