Tetrahydrofurfuryl propionate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T1016-25ML T1016-500ML
TCIAT1016-25MLEA 46.2 EUR
TCIAT1016-25ML TCIAT1016-500ML
Tetrahydrofurfuryl propionate ≥97.0% (by GC)
Tetrahydrofurfuryl propionate
Formule: C₈H₁₄O₃
Kookpunt: 205°C
Vlampunt: 92°C
MDL Nummer: MFCD00047080
CAS nummer: 637-65-0
EINECS: 211-297-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec5ed12289baff478c898e4ecaa4d359c6f07ecd&vid=ec5ed12289baff478c898e4ecaa4d359c6f07ecd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR