1,1-Dimethylbiguanide hydrochloride
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride
J63361.06
J63361.14
J63361.22
J63361.06EA
41.6
EUR
J63361.06
J63361.14
J63361.22
1,1-Dimethylbiguanide hydrochloride
1,1-Dimethylbiguanide hydrochloride
CAS No.: 1115-70-4
Molecular Formula: C4H11N5.HCl
Formula Weight: 165.62
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 218-226°
Merck Reference: 14,5938
MDL No.: MFCD00012582
An antidiabetic agent
Formule:
C₄H₁₂ClN₅ Smeltpunt: 218…226 °C |
MDL Nummer:
MFCD00012582 CAS nummer: 1115-70-4 EINECS: 214-230-6 Merck Index: 14,5938 |