Pilocarpine hydrochloride
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (+)-Pilocarpine hydrochloride
14487-500 14487-1 14487-5 14487-10
CAYM14487-500EA 57.1 EUR
CAYM14487-500 CAYM14487-1 CAYM14487-5 CAYM14487-10
Pilocarpine hydrochloride
Pilocarpine hydrochloride
Formule: C₁₁H₁₇ClN₂O₂
MDL Nummer: MFCD00012722
CAS nummer: 54-71-7
EINECS: 200-212-5