Bathocuproin sulphonate disodium salt for determination of Cu in blood

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B0985-1G B0985-100MG
TCIAB0985-1GEA 260 EUR
TCIAB0985-1G TCIAB0985-100MG
Bathocuproin sulphonate disodium salt for determination of Cu in blood
Bathocuproin sulphonate disodium salt
Formule: C₂₆H₁₈N₂Na₂O₆S₂
MDL Nummer: MFCD00011679
CAS nummer: 52698-84-7
EINECS: 258-111-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95afa627dd77e9701f605b01d535e11b17dac945&vid=886dc9a0b9a9d81e6377da33ae0c95b88c207d37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Absorbance of Cu complex min. 12,000
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR