4-(4-Fluorobenzoyl)piperidine hydrochloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CD09967.250 CD09967.1
MAYBCD09967.250EA 0 EUR
MAYBCD09967.250 MAYBCD09967.1
4-(4-Fluorobenzoyl)piperidine hydrochloride
4-(4-Fluorobenzoyl)piperidine hydrochloride
Formule: C₁₂H₁₅ClFNO
MDL Nummer: MFCD00044912
CAS nummer: 25519-78-2
EINECS: 247-070-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR