1-Pyreneboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Pyrene-1-boronic acid, Pyren-1-ylboronic acid

P1625-1G P1625-25G P1625-5G
TCIAP1625-1GEA 27.5 EUR
TCIAP1625-1G TCIAP1625-25G TCIAP1625-5G
1-Pyreneboronic acid (contains varying amounts of Anhydride)
1-Pyreneboronic acid
Formule: C₁₆H₁₁BO₂
MDL Nummer: MFCD04974062
CAS nummer: 164461-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105a4ca15c7caad467469e9449ae8ebf596063b7&vid=105a4ca15c7caad467469e9449ae8ebf596063b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 95.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR