4,6-Dichloro-2-(methylthio)pyrimidine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D2533-25G D2533-5G
TCIAD2533-25GEA 59.4 EUR
TCIAD2533-25G TCIAD2533-5G
4,6-Dichloro-2-(methylthio)pyrimidine ≥98.0% (by GC)
4,6-Dichloro-2-(methylthio)pyrimidine
Formule: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Kookpunt: 136°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006086
CAS nummer: 6299-25-8
EINECS: 228-577-6
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a044a1f502ac0c85cad0e778dbdca5637f49a780&vid=a044a1f502ac0c85cad0e778dbdca5637f49a780&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR