Tributyl(5-fluoro-2-methoxyphenyl)stannane
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
SP00185.1
SP00185.10
SP00185.50
MAYBSP00185.1EA
0
EUR
MAYBSP00185.1
MAYBSP00185.10
MAYBSP00185.50
Tributyl(5-fluoro-2-methoxyphenyl)stannane
Tributyl(5-fluoro-2-methoxyphenyl)stannane
Formule:
C₁₉H₃₃FOSn |
CAS nummer:
223432-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...