Picolinamidine hydrochloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Picolinimidamide hydrochloride, 2-Amidinopyridine hydrochloride

RF01042DA RF01042EA
MAYBRF01042DAEA 0 EUR
MAYBRF01042DA MAYBRF01042EA
Picolinamidine hydrochloride
Picolinamidine hydrochloride
Formule: C₆H₈ClN₃
MDL Nummer: MFCD00052271
CAS nummer: 51285-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR