2-(Methylthio)thiazole ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M1712-25G M1712-5G
TCIAM1712-25GEA 119 EUR
TCIAM1712-25G TCIAM1712-5G
2-(Methylthio)thiazole ≥98.0% (by GC)
2-(Methylthio)thiazole
Formule: C₄H₅NS₂
Kookpunt: 62°C
Vlampunt: 91°C
MDL Nummer: MFCD00858981
CAS nummer: 5053-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eb4d247e3e3f50e67a79c77f4ceb28b2f0bfbbc&vid=2eb4d247e3e3f50e67a79c77f4ceb28b2f0bfbbc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %