1,2-Ethanedisulphonic acid dihydrate 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethane-1,2-disulphonic acid dihydrate
384800050
384800250
ACRO384800050EA
55.2
EUR
ACRO384800050
ACRO384800250
1,2-Ethanedisulphonic acid dihydrate 98%
1,2-Ethanedisulphonic acid dihydrate
Formule:
C₂H₆O₆S₂·2H₂O Smeltpunt: 109…113 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD26793415 CAS nummer: 5982-56-9 EINECS: 203-732-0 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | Off-white to beige to grey to light brown Powder or crystals |
Infrared spectrum | Conforms |
Water | 14 to 19 % (K.F.) |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...