N-Boc-D-valine ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Boc-D-Val-OH
L09193.03
L09193.06
L09193.03EA
25.1
EUR
L09193.03
L09193.06
N-Boc-D-valine ≥98%
N-Boc-D-valine
Purity: 98+%
CAS No.: 22838-58-0
Molecular Formula: C10H19NO4
Formula Weight: 217.27
Storage Temperature: 2°C to 8°C
Melting Point: 76-80°
Beilstein Reference: 2050408
Optical Rotation: +6.3° (c=1 in acetic acid)
MDL No.: MFCD00038282
Formule:
C₁₀H₁₉NO₄ Smeltpunt: 76…80 °C |
MDL Nummer:
MFCD00038282 CAS nummer: 22838-58-0 |