N-Boc-L-valine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Boc-Val-OH, BOC-L-Valine

4000426.0100 4000426.1000 4000426.0500
BACE4000426.0100EA 0 EUR
BACE4000426.0100 BACE4000426.1000 BACE4000426.0500
N-Boc-L-valine
N-Boc-L-valine
Formule: C₁₀H₁₉NO₄
MW: 217.3 g/mol
MDL Nummer: MFCD00065605
CAS nummer: 13734-41-3
EINECS: 237-307-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR