4-Bromo-N,N-diphenylaniline ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B2633-25G
B2633-5G
TCIAB2633-25GEA
192
EUR
TCIAB2633-25G
TCIAB2633-5G
4-Bromo-N,N-diphenylaniline ≥97.0% (by GC)
4-Bromo-N,N-diphenylaniline
Formule:
C₁₈H₁₄BrN Kookpunt: 185°C |
MDL Nummer:
MFCD01851266 CAS nummer: 36809-26-4 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...