Tetrabutylammonium p-nitrophenoxide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2669-25G T2669-5G
TCIAT2669-25GEA 104 EUR
TCIAT2669-25G TCIAT2669-5G
Tetrabutylammonium p-nitrophenoxide ≥98.0% (by HPLC, titration analysis)
Tetrabutylammonium p-nitrophenoxide
Formule: C₂₂H₄₀N₂O₃
CAS nummer: 3002-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ce664bcf3cb3b22a6964cb51b636c830b1517c&vid=27ce664bcf3cb3b22a6964cb51b636c830b1517c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Melting point 143.0 to 147.0 deg-C
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR