N-Boc-L-phenylalaninol

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-(-)-BOC-Phenylalaninol, Boc-Phenylalaninol, (S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate

4010424.0005 4010424.0025
BACE4010424.0005EA 0 EUR
BACE4010424.0005 BACE4010424.0025
N-Boc-L-phenylalaninol
N-Boc-L-phenylalaninol
Formule: C₁₄H₂₁NO₃
MDL Nummer: MFCD00076976
CAS nummer: 66605-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR