2-Amino-4-methylbenzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00794DA TL00794EA
MAYBTL00794DAEA 0 EUR
MAYBTL00794DA MAYBTL00794EA
2-Amino-4-methylbenzamide
2-Amino-4-methylbenzamide
Formule: C₈H₁₀N₂O
MDL Nummer: MFCD00221474
CAS nummer: 39549-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR