1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1403-25G B1403-5G
TCIAB1403-25GEA 269 EUR
TCIAB1403-25G TCIAB1403-5G
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98.0% (by GC)
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Formule: C₁₂H₆F₁₂O₂
Kookpunt: 100°C
Vlampunt: 97°C
MDL Nummer: MFCD00042090
CAS nummer: 802-93-7
EINECS: 212-354-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=139770db9889cdd61b1cee72f303cfd73da2e641&vid=139770db9889cdd61b1cee72f303cfd73da2e641&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %