2-Phenoxytetrahydropyran ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0853-25ML P0853-5ML
TCIAP0853-25MLEA 148 EUR
TCIAP0853-25ML TCIAP0853-5ML
2-Phenoxytetrahydropyran ≥90.0% (by GC)
2-Phenoxytetrahydropyran
Formule: C₁₁H₁₄O₂
MDL Nummer: MFCD00059801
CAS nummer: 4203-50-3
EINECS: 224-114-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27013ea78717252fb9d10b2ccd8233ffc560bb7a&vid=27013ea78717252fb9d10b2ccd8233ffc560bb7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR