2,6-Dichloro-3-cyano-4-picoline ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine
D3346-25G D3346-5G
TCIAD3346-25GEA 78.1 EUR
TCIAD3346-25G TCIAD3346-5G
2,6-Dichloro-3-cyano-4-picoline ≥98.0% (by GC)
2,6-Dichloro-3-cyano-4-picoline
Formule: C₇H₄Cl₂N₂
Kookpunt: 118°C
MDL Nummer: MFCD00052633
CAS nummer: 875-35-4
EINECS: 212-873-7
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=830e36b25e79324e44445e95c58da12c588acdcf&vid=830e36b25e79324e44445e95c58da12c588acdcf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 109.0 to 113.0 deg-C