2-(4-Chlorobenzoyl)pyridine ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
(4-Chlorophenyl)(pyridin-2-yl)methanone
C2647-25G
C2647-5G
TCIAC2647-25GEA
63.8
EUR
TCIAC2647-25G
TCIAC2647-5G
2-(4-Chlorobenzoyl)pyridine ≥97.0% (by GC, titration analysis)
2-(4-Chlorobenzoyl)pyridine
Formule:
C₁₂H₈ClNO Kookpunt: 160°C |
MDL Nummer:
MFCD02930888 CAS nummer: 6318-51-0 EINECS: 228-662-8 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...