4-Morpholino-3-nitrobenzaldehyde

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

RH01168.1 RH01168.10
MAYBRH01168.1EA 0 EUR
MAYBRH01168.1 MAYBRH01168.10
4-Morpholino-3-nitrobenzaldehyde
4-Morpholino-3-nitrobenzaldehyde
Formule: C₁₁H₁₂N₂O₄
CAS nummer: 300541-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR