4-[5-Chloro-2-nitro-4-(trifluoromethyl)phenyl]morpholine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB02404.250 BTB02404.1
MAYBBTB02404.250EA 0 EUR
MAYBBTB02404.250 MAYBBTB02404.1
4-[5-Chloro-2-nitro-4-(trifluoromethyl)phenyl]morpholine
4-[5-Chloro-2-nitro-4-(trifluoromethyl)phenyl]morpholine
Formule: C₁₁H₁₀ClF₃N₂O₃
CAS nummer: 35289-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR