Methyl-1,2,5-trimethylpyrrole-3-carboxylate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SP00005.1 SP00005.10
MAYBSP00005.1EA 0 EUR
MAYBSP00005.1 MAYBSP00005.10
Methyl-1,2,5-trimethylpyrrole-3-carboxylate
Methyl-1,2,5-trimethylpyrrole-3-carboxylate
Formule: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Nummer: MFCD00203867
CAS nummer: 14186-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR