1-Isopropyl-4-piperidone ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0566-25G I0566-5G
TCIAI0566-25GEA 77 EUR
TCIAI0566-25G TCIAI0566-5G
1-Isopropyl-4-piperidone ≥97.0% (by titrimetric analysis)
1-Isopropyl-4-piperidone
Formule: C₈H₁₅NO
Kookpunt: 101°C
MDL Nummer: MFCD00038035
CAS nummer: 5355-68-0
EINECS: 226-339-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0ec5b444bb7f9336fa1d23f2cd6c6ec0f369fc5&vid=b0ec5b444bb7f9336fa1d23f2cd6c6ec0f369fc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR