(R)-(+)-2-Hydroxy-3-phenylpropionic acid

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

4013938.0001 4013938.0005
BACE4013938.0001EA 0 EUR
BACE4013938.0001 BACE4013938.0005
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Formule: C₉H₁₀O₃
MDL Nummer: MFCD00078062
CAS nummer: 7326-19-4
EINECS: 230-803-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR