1,4-Diiodo-2,5-dimethoxybenzene
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB13697.1
BTB13697.10
MAYBBTB13697.1EA
0
EUR
MAYBBTB13697.1
MAYBBTB13697.10
1,4-Diiodo-2,5-dimethoxybenzene
1,4-Diiodo-2,5-dimethoxybenzene
Formule:
C₈H₈I₂O₂ MW: 389.9569 g/mol |
MDL Nummer:
MFCD00461369 CAS nummer: 51560-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...