1-Bromo-3-isopropoxybenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B2198-25G B2198-5G
TCIAB2198-25GEA 77 EUR
TCIAB2198-25G TCIAB2198-5G
1-Bromo-3-isopropoxybenzene ≥98.0% (by GC)
1-Bromo-3-isopropoxybenzene
Formule: C₉H₁₁BrO
Vlampunt: 109°C
MDL Nummer: MFCD00070756
CAS nummer: 131738-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=864d686502b61bbdab2e654b0f394916e3342195&vid=864d686502b61bbdab2e654b0f394916e3342195&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR