4-Nonylbiphenyl

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB10233.1 BTB10233.10
MAYBBTB10233.1EA 0 EUR
MAYBBTB10233.1 MAYBBTB10233.10
4-Nonylbiphenyl
4-Nonylbiphenyl
Formule: C₂₁H₂₈
MDL Nummer: MFCD00100666
CAS nummer: 93972-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR