2-(2-Bromoethyl)-1,3-dioxane ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L00958.09
L00958.18
L00958.09EA
35.3
EUR
L00958.09
L00958.18
2-(2-Bromoethyl)-1,3-dioxane ≥98%
2-(2-Bromoethyl)-1,3-dioxane
Purity: 98%
CAS No.: 33884-43-4
Molecular Formula: C6H11BrO2
Formula Weight: 195.06
Storage Temperature: 2°C to 8°C
Flash Point: 96°(204°F)
Refractive Index: 1.4820
Boiling Point: 67-70°/3mm
Beilstein Reference: 1421628
MDL No.: MFCD00006567
Formule:
C₆H₁₁BrO₂ MW: 195.06 g/mol Kookpunt: 67…70 °C (3 mmHg) Dichtheid: 1.434 Vlampunt: 96 °C (204 °F) |
MDL Nummer:
MFCD00006567 CAS nummer: 33884-43-4 EINECS: 251-716-7 |