(S)-(+)-Ethyl-3-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate
N0679-25G
N0679-5G
TCIAN0679-25GEA
479
EUR
TCIAN0679-25G
TCIAN0679-5G
(S)-(+)-Ethyl-3-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
(S)-(+)-Ethyl-3-piperidinecarboxylate
Formule:
C₈H₁₅NO₂ MW: 157.21 g/mol |
MDL Nummer:
MFCD00792499 CAS nummer: 37675-18-6 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | 0.0 to +2.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...