2,3,5-Trifluoropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T2778-25G
T2778-5G
TCIAT2778-25GEA
334
EUR
TCIAT2778-25G
TCIAT2778-5G
2,3,5-Trifluoropyridine ≥98.0% (by GC)
2,3,5-Trifluoropyridine
Formule:
C₅H₂F₃N Kookpunt: 102°C Dichtheid: 1,499 g/cm³ (20 °C) Vlampunt: 30°C |
MDL Nummer:
MFCD03001162 CAS nummer: 76469-41-5 UN: 1993 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |