4'-Hydroxy-3'-methoxyacetophenone
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
11976-5 11976-10 11976-1
CAYM11976-5EA 269 EUR
CAYM11976-5 CAYM11976-10 CAYM11976-1
4'-Hydroxy-3'-methoxyacetophenone
4'-Hydroxy-3'-methoxyacetophenone
Formule: C₉H₁₀O₃
Kookpunt: 295 °C (1013 hPa)
Smeltpunt: 115 °C
MDL Nummer: MFCD00008747
CAS nummer: 498-02-2
EINECS: 207-854-5
Merck Index: 13,00746