3,4-Dimethyl-o-phenylenediamine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB06728.1 BTB06728.10 BTB06728.50
MAYBBTB06728.1EA 0 EUR
MAYBBTB06728.1 MAYBBTB06728.10 MAYBBTB06728.50
3,4-Dimethyl-o-phenylenediamine
3,4-Dimethyl-o-phenylenediamine
Formule: C₈H₁₂N₂
Kookpunt: 287 °C (1013 hPa)
Smeltpunt: 89 °C
MDL Nummer: MFCD00051709
CAS nummer: 41927-01-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR