4,5-Dichloro-o-phenylenediamine
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine
BTB07389.1 BTB07389.10 BTB07389.50
MAYBBTB07389.1EA 0 EUR
MAYBBTB07389.1 MAYBBTB07389.10 MAYBBTB07389.50
4,5-Dichloro-o-phenylenediamine
4,5-Dichloro-o-phenylenediamine
Formule: C₆H₆Cl₂N₂
Kookpunt: 305 °C (1013 hPa)
Smeltpunt: 163 °C
MDL Nummer: MFCD00007723
CAS nummer: 5348-42-5
EINECS: 226-305-0
UN: 2811
ADR: 6.1,III