4,5-Dimethyl-o-phenylenediamine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene

TL00793.1 TL00793.10 TL00793.50
MAYBTL00793.1EA 0 EUR
MAYBTL00793.1 MAYBTL00793.10 MAYBTL00793.50
4,5-Dimethyl-o-phenylenediamine
4,5-Dimethyl-o-phenylenediamine
Formule: C₈H₁₂N₂
MDL Nummer: MFCD00007729
CAS nummer: 3171-45-7
EINECS: 221-635-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR