4-Bromo-3-fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3835-5G B3835-1G
TCIAB3835-5GEA 244 EUR
TCIAB3835-5G TCIAB3835-1G
4-Bromo-3-fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-Bromo-3-fluorobenzenesulphonyl chloride
Formule: C₆H₃BrClFO₂S
Kookpunt: 217 - 218 °C (1013 hPa)
Dichtheid: 1,85 g/cm³ (20 °C)
MDL Nummer: MFCD03094150
CAS nummer: 351003-51-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=279c9a0a005d36d6da46fd3076e9f8c9998c4418&vid=279c9a0a005d36d6da46fd3076e9f8c9998c4418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR