Benzethonium chloride 0.004 mol/l, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hyamine
7170.1000EA 37.05 EUR
7170.1000 7170.2500
Benzethonium chloride 0.004 mol/l, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Benzethonium chloride
Formule: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
MDL Nummer: MFCD00011742
CAS nummer: 121-54-0
EINECS: 204-479-9
Merck Index: 13,01074
Order Now
Specification Test Results
Titer 0.0039 - 0.0041 mol/l