Azomethine H sodium salt 97,0 Titrimetric Spectrophotometric Reagent for B

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt, Azomethine H, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid monosodium salt, Azomethine-H monosodium salt

A5016-25G A5016-5G
TCIAA5016-25GEA 164 EUR
TCIAA5016-25G TCIAA5016-5G
Azomethine H sodium salt 97,0 Titrimetric Spectrophotometric Reagent for B
Azomethine H sodium salt
Formule: C₁₇H₁₂NNaO₈S₂
MW: 445.41 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042050
CAS nummer: 5941-07-1
EINECS: 227-698-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2750acdb3a146e587e23931932c43365067684c6&vid=67ef58aa7755b3e64e1d2a46292c95842d8f999a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR