Methylprednisolone

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 6α-Methylprednisolone

15013-500 15013-5 15013-1
CAYM15013-500EA 111 EUR
CAYM15013-500 CAYM15013-5 CAYM15013-1
Methylprednisolone
Methylprednisolone
Formule: C₂₂H₃₀O₅
MW: 374.48 g/mol
Smeltpunt: 230 °C
MDL Nummer: MFCD00010591
CAS nummer: 83-43-2
EINECS: 201-476-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR