N-Methyl-4-nitroaniline
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01115.1
SB01115.10
SB01115.50
MAYBSB01115.1EA
0
EUR
MAYBSB01115.1
MAYBSB01115.10
MAYBSB01115.50
N-Methyl-4-nitroaniline
N-Methyl-4-nitroaniline
Formule:
C₇H₈N₂O₂ MW: 152.15 g/mol Kookpunt: 291 Smeltpunt: 148…149 °C Dichtheid: 1.201 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007305 CAS nummer: 100-15-2 EINECS: 202-823-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...