Benzyltrimethylammonium hydroxide 40% in aqueous solution
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
8.45025.5000 8.45025.0100
8.45025.5000EA 0 EUR
8.45025.5000 8.45025.0100
Benzyltrimethylammonium hydroxide 40% in aqueous solution
Benzyltrimethylammonium hydroxide
Formule: C₁₀H₁₇NO
MW: 167.25
MDL Nummer: MFCD00008281
CAS nummer: 100-85-6
EINECS: 202-895-5
UN: 3267
ADR: 8 II