5-Nitroso-8-quinolinol
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13856-10 13856-50 13856-100
CAYM13856-10EA 57.1 EUR
CAYM13856-10 CAYM13856-50 CAYM13856-100
5-Nitroso-8-quinolinol
5-Nitroso-8-quinolinol
Formule: C₉H₆N₂O₂
Smeltpunt: 245 °C
MDL Nummer: MFCD00041862
CAS nummer: 3565-26-2
EINECS: 222-650-6
UN: 2811
ADR: 6.1,III