3-Furoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Furan-3-carboxylic acid, 3-Furancarboxylic acid

F0304-25G F0304-5G
TCIAF0304-25GEA 214 EUR
TCIAF0304-25G TCIAF0304-5G
3-Furoic acid ≥98.0% (by titrimetric analysis)
3-Furoic acid
Formule: C₅H₄O₃
MDL Nummer: MFCD00005350
CAS nummer: 488-93-7
EINECS: 207-689-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8271384370ef409b54740fdeafcd76b77089bd09&vid=8271384370ef409b54740fdeafcd76b77089bd09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 124.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR