Procaine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-(Diethylamino)ethyl 4-Aminobenzoate, 4-Aminobenzoic acid 2-(Diethylamino)ethyl ester, 2-(Diethylamino)ethyl 4-aminobenzoate

P1961-25G P1961-5G
TCIAP1961-25GEA 73.7 EUR
TCIAP1961-25G TCIAP1961-5G
Procaine ≥98.0% (by HPLC, titration analysis)
Procaine
Formule: C₁₃H₂₀N₂O₂
MDL Nummer: MFCD00007893
CAS nummer: 59-46-1
EINECS: 200-426-9
Merck Index: 14,07757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adeb0588ab34d88c2207af628b7b955f21b62b29&vid=adeb0588ab34d88c2207af628b7b955f21b62b29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR