5-Methoxy-1-tetralone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2297-5G M2297-25G
TCIAM2297-5GEA 83.6 EUR
TCIAM2297-5G TCIAM2297-25G
5-Methoxy-1-tetralone ≥98.0% (by GC)
5-Methoxy-1-tetralone
Formule: C₁₁H₁₂O₂
MDL Nummer: MFCD00001692
CAS nummer: 33892-75-0
EINECS: 251-723-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0032d01c427219957635fd599644ce34743bdee3&vid=0032d01c427219957635fd599644ce34743bdee3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR