Methyltributylammonium chloride ∼75% ≥75%

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2873-500ML T2873-100ML
TCIAT2873-500MLEA 47.3 EUR
TCIAT2873-500ML TCIAT2873-100ML
Methyltributylammonium chloride ∼75% ≥75%
Methyltributylammonium chloride
Formule: C₁₃H₃₀ClN
MDL Nummer: MFCD00011847
CAS nummer: 56375-79-2
EINECS: 260-135-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ffb9c62f126d7249c143f16bdf2af185a07e5f2&vid=68aca6fabac62204c6df3d3287a90fa7c0a6491b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Concentration (Nonaqueous Titration) 73.0 to 78.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR