Mounting/reducer clips for tubes
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
552 550 554 551 553
BOCH552EA 174 EUR
BOCH552 BOCH550 BOCH554 BOCH551 BOCH553
Mounting/reducer clips for tubes
Klemmen Klemmen voor algemeen gebruik
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.